COMH4211A99E Combi-Blocks, Inc.

5-Hydroxy-2-(2-methoxy-ethoxy)-benzoic acid methyl ester

  • MDL number MFCD31618916
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0451
Availability details for
Quantity
  • Availability
  • Add to Cart