COMH4211A99F Combi-Blocks, Inc.

1-[3-(2,2-Difluoroethoxy)phenyl]-n-methylmethanamine

  • MDL number MFCD12807341
  • Cas Number : 1183766-38-2
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0452

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0452
Availability details for
Quantity
  • Availability
  • Add to Cart