COMH4211A9A1 Combi-Blocks, Inc.

L-Leucinamide, n-[(phenylmethoxy)carbonyl]-beta-alanyl-

  • MDL number MFCD00136506
  • Cas Number : 197237-13-1
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0457
Availability details for
Quantity
  • Availability
  • Add to Cart