COMH4211A9A2 Combi-Blocks, Inc.

4-Amino-2-fluoro-n-(2-fluoroethyl)benzamide

  • MDL number MFCD31583032
  • Cas Number : 1857178-98-3
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0459
Availability details for
Quantity
  • Availability
  • Add to Cart