COMH4211A9A3 Combi-Blocks, Inc.

Benzenesulfonic acid, 4-methyl-, methylazanyl ester

  • MDL number MFCD24873142
  • Cas Number : 25370-97-2
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0461

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0461
Availability details for
Quantity
  • Availability
  • Add to Cart