COMH4211A9A4 Combi-Blocks, Inc.

3-Bromo-4-(methylsulfonyl)benzenamine

  • MDL number MFCD16085639
  • Cas Number : 955879-83-1
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0466
Availability details for
Quantity
  • Availability
  • Add to Cart