COMH4211A9A6 Combi-Blocks, Inc.

2-(tert-Butylamino)-3'-bromopropiophenone hydrochloride

  • MDL number MFCD01749197
  • Cas Number : 1049718-43-5
  • Molecular Weight : 320.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0468
Availability details for
Quantity
  • Availability
  • Add to Cart