COMH4211A9A7 Combi-Blocks, Inc.

4-(4-Hydroxymethyl-3-trifluoromethyl-phenyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583156
  • Cas Number : 1240970-32-4
  • Molecular Weight : 359.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0470
Availability details for
Quantity
  • Availability
  • Add to Cart