COMH4211A9AA Combi-Blocks, Inc.

1-(2,5-Dimethylbenzoyl)piperidine-4-carboxylic acid

  • MDL number MFCD09802928
  • Cas Number : 1016715-89-1
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0476
Availability details for
Quantity
  • Availability
  • Add to Cart