COMH4211A9AB Combi-Blocks, Inc.

2-Azabicyclo[2.2.2]octane-3,5-dione

  • MDL number MFCD13178731
  • Cas Number : 596795-04-9
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0477

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0477
Availability details for
Quantity
  • Availability
  • Add to Cart