COMH4211A9AC Combi-Blocks, Inc.

1-(4-Fluoro-2-methylphenyl)propan-1-amine

  • MDL number MFCD17222793
  • Cas Number : 1270440-21-5
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0480
Availability details for
Quantity
  • Availability
  • Add to Cart