COMH4211A9AD Combi-Blocks, Inc.

tert-Butyl 4-(4-bromo-2-fluorobenzoyl)piperazine-1-carboxylate

  • MDL number MFCD17872926
  • Cas Number : 1223433-71-3
  • Molecular Weight : 387.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0481
Availability details for
Quantity
  • Availability
  • Add to Cart