COMH4211A9AE Combi-Blocks, Inc.

3-Fluoro-5-methoxy-4-nitrobenzonitrile

  • MDL number MFCD23701681
  • Cas Number : 1137869-92-1
  • Molecular Weight : 196.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0482
Availability details for
Quantity
  • Availability
  • Add to Cart