COMH4211A9AF Combi-Blocks, Inc.

1-[4-(2-Methoxyethoxy)phenyl]propan-2-one

  • MDL number MFCD24220067
  • Cas Number : 91517-62-3
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0484
Availability details for
Quantity
  • Availability
  • Add to Cart