COMH4211A9B0 Combi-Blocks, Inc.

5,5-Difluoro-1,3-dimethyl-7-(3-oxo-3-(prop-2-yn-1-ylamino)propyl)-5h-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide

  • MDL number MFCD28400774
  • Cas Number : 302795-84-2
  • Molecular Weight : 329.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0487
Availability details for
Quantity
  • Availability
  • Add to Cart