COMH4211A9B1 Combi-Blocks, Inc.

tert-Butyl cyclopropylsulfonylcarbamate

  • MDL number MFCD16660018
  • Cas Number : 681808-26-4
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0489

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0489
Availability details for
Quantity
  • Availability
  • Add to Cart