COMH4211A9B2 Combi-Blocks, Inc.

[4-(Oxetan-3-yloxy)-phenyl]-methanol

  • MDL number MFCD21848554
  • Cas Number : 1780449-65-1
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0493
Availability details for
Quantity
  • Availability
  • Add to Cart