COMH4211A9B4 Combi-Blocks, Inc.

4-(4-M-Tolyl-piperazin-1-yl)-butyric acid dihydrochloride

  • MDL number MFCD31583323
  • Cas Number : 2270905-37-6
  • Molecular Weight : 335.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0496
Availability details for
Quantity
  • Availability
  • Add to Cart