COMH4211A9B5 Combi-Blocks, Inc.

1-[3-Bromo-4-(propan-2-yloxy)phenyl]methanamine

  • MDL number MFCD17284199
  • Cas Number : 1339365-08-0
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0497
Availability details for
Quantity
  • Availability
  • Add to Cart