COMH4211A9BB Combi-Blocks, Inc.

[(4-Chloro-2-fluorophenyl)methyl](ethyl)amine

  • MDL number MFCD11212422
  • Cas Number : 1094511-87-1
  • Molecular Weight : 187.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0511

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0511
Availability details for
Quantity
  • Availability
  • Add to Cart