COMH4211A9BC Combi-Blocks, Inc.

5-Bromo-2-dimethylamino-nicotinic acid tert-butyl ester

  • MDL number MFCD31557892
  • Cas Number : 2206609-32-5
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0513
Availability details for
Quantity
  • Availability
  • Add to Cart