COMH4211A9BD Combi-Blocks, Inc.

2-(4-Hydroxy-pyrazol-1-yl)-2-methyl-propionic acid tert-butyl ester

  • MDL number MFCD31583107
  • Cas Number : 2270907-01-0
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0514
Availability details for
Quantity
  • Availability
  • Add to Cart