COMH4211A9BE Combi-Blocks, Inc.

6-Chloro-pyridine-2-carboxylic acid diethylamide

  • MDL number MFCD16698134
  • Cas Number : 227453-44-3
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-N,N-diethyl-2-pyridinecarboxamide
Inchi Code 1S/C10H13ClN2O/c1-3-13(4-2)10(14)8-6-5-7-9(11)12-8/h5-7H,3-4H2,1-2H3
Inchi Key FGCMQANIPVXFJM-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0515

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0515
Availability details for
Quantity
  • Availability
  • Add to Cart