COMH4211A9BF Combi-Blocks, Inc.

4-Chloro-2-iodo-n-methoxy-n-methylbenzamide

  • MDL number MFCD11847518
  • Cas Number : 1262282-26-7
  • Molecular Weight : 325.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0517
Availability details for
Quantity
  • Availability
  • Add to Cart