COMH4211A9C0 Combi-Blocks, Inc.

tert-Butyl ((3-chloropyrazin-2-yl)methyl)carbamate

  • MDL number MFCD28043784
  • Cas Number : 1823551-51-4
  • Molecular Weight : 243.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0519

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0519
Availability details for
Quantity
  • Availability
  • Add to Cart