COMH4211A9C1 Combi-Blocks, Inc.

4-Amino-2-(2,2,2-trifluoro-ethoxy)-benzoic acid methyl ester

  • MDL number MFCD31557918
  • Cas Number : 1970385-57-9
  • Molecular Weight : 249.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0520

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0520
Availability details for
Quantity
  • Availability
  • Add to Cart