COMH4211A9C2 Combi-Blocks, Inc.

Cyclopropanesulfonic acid [3-bromo-5-(pyrrolidine-1-carbonyl)-phenyl]-amide

  • MDL number MFCD31613988
  • Cas Number : 2270906-99-3
  • Molecular Weight : 373.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0521
Availability details for
Quantity
  • Availability
  • Add to Cart