COMH4211A9C3 Combi-Blocks, Inc.

3'-Chloro-3-methyl-biphenyl-4-carboxylic acid

  • MDL number MFCD31561445
  • Molecular Weight : 246.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0527

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0527
Availability details for
Quantity
  • Availability
  • Add to Cart