COMH4211A9C4 Combi-Blocks, Inc.

4-(3-Bromo-phenoxymethyl)-1-cyclopropylmethyl-piperidine

  • MDL number MFCD31539563
  • Cas Number : 189618-25-5
  • Molecular Weight : 324.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0528

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0528
Availability details for
Quantity
  • Availability
  • Add to Cart