COMH4211A9C5 Combi-Blocks, Inc.

2-Iodo-4-thiophen-2-yl-pyridine

  • MDL number MFCD31613893
  • Cas Number : 1334675-37-4
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0530
Availability details for
Quantity
  • Availability
  • Add to Cart