COMH4211A9C7 Combi-Blocks, Inc.

(1-Methanesulfonylpyrrolidin-2-yl)methanol

  • MDL number MFCD11897235
  • Cas Number : 1176673-70-3
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0532
Availability details for
Quantity
  • Availability
  • Add to Cart