COMH4211A9C9 Combi-Blocks, Inc.

Methyl 3-iodopropanoate

  • MDL number MFCD24458739
  • Cas Number : 5029-66-3
  • Molecular Weight : 214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-iodopropanoate
Inchi Code 1S/C4H7IO2/c1-7-4(6)2-3-5/h2-3H2,1H3
Inchi Key YIYZNTTWHIYCOO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0541
Availability details for
Quantity
  • Availability
  • Add to Cart