COMH4211A9CB Combi-Blocks, Inc.

3-Bromo-4-(2-iodophenoxy)-benzonitrile

  • MDL number MFCD25801529
  • Cas Number : 1325219-94-0
  • Molecular Weight : 400.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0546
Availability details for
Quantity
  • Availability
  • Add to Cart