COMH4211A9CC Combi-Blocks, Inc.

1-(2-Bromo-5-ethoxy-4-methyl-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31557812
  • Cas Number : 2206608-62-8
  • Molecular Weight : 294.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0548
Availability details for
Quantity
  • Availability
  • Add to Cart