COMH4211A9CD Combi-Blocks, Inc.

4-(Benzyloxy)-2-tert-butylphenol

  • MDL number MFCD17012739
  • Cas Number : 3421-95-2
  • Molecular Weight : 256.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(benzyloxy)-2-tert-butylphenol
Inchi Code 1S/C17H20O2/c1-17(2,3)15-11-14(9-10-16(15)18)19-12-13-7-5-4-6-8-13/h4-11,18H,12H2,1-3H3
Inchi Key PFBOAJKGEXKOBX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0549

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0549
Availability details for
Quantity
  • Availability
  • Add to Cart