COMH4211A9D1 Combi-Blocks, Inc.

Benzene, 2-bromo-1-[(1-methylethyl)sulfonyl]-4-nitro-

  • MDL number MFCD27931836
  • Cas Number : 900174-33-6
  • Molecular Weight : 308.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0554
Availability details for
Quantity
  • Availability
  • Add to Cart