COMH4211A9D2 Combi-Blocks, Inc.

6-(4-Formyl-phenoxy)-nicotinamide

  • MDL number MFCD30549207
  • Cas Number : 676494-70-5
  • Molecular Weight : 242.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(4-formylphenoxy)nicotinamide
Inchi Code 1S/C13H10N2O3/c14-13(17)10-3-6-12(15-7-10)18-11-4-1-9(8-16)2-5-11/h1-8H,(H2,14,17)
Inchi Key WMRYYTSBKKACAN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0555

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0555
Availability details for
Quantity
  • Availability
  • Add to Cart