COMH4211A9D3 Combi-Blocks, Inc.

6-Bromo-1-(cyclopropylmethyl)-1h-pyrrolo[3,2-b]pyridine

  • MDL number MFCD31613918
  • Cas Number : 1801905-66-7
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0560
Availability details for
Quantity
  • Availability
  • Add to Cart