COMH4211A9D4 Combi-Blocks, Inc.

4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazole-1-carboxylic acid ethylamide

  • MDL number MFCD31583313
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0561
Availability details for
Quantity
  • Availability
  • Add to Cart