COMH4211A9D6 Combi-Blocks, Inc.

4-Amino-2-(2-methylpropoxy)benzamide

  • MDL number MFCD19640754
  • Cas Number : 99981-76-7
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0568

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0568
Availability details for
Quantity
  • Availability
  • Add to Cart