COMH4211A9D7 Combi-Blocks, Inc.

(3-Butyl-5-methyl-isoxazol-4-yl)-methanol

  • MDL number MFCD28362967
  • Cas Number : 1239465-21-4
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0571
Availability details for
Quantity
  • Availability
  • Add to Cart