COMH4211A9DB Combi-Blocks, Inc.

2-Isobutoxy-5-methyl-benzylamine

  • MDL number MFCD18331101
  • Cas Number : 1344007-38-0
  • Molecular Weight : 193.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0579
Availability details for
Quantity
  • Availability
  • Add to Cart