COMH4211A9DC Combi-Blocks, Inc.

Trans 3-(4-tert-butoxycarbonylamino-cyclohexyl)-2-methyl-acrylic acid ethyl ester

  • MDL number MFCD31579304
  • Cas Number : 2365390-53-8
  • Molecular Weight : 311.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0580

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0580
Availability details for
Quantity
  • Availability
  • Add to Cart