COMH4211A9DD Combi-Blocks, Inc.

4-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)aniline

  • MDL number MFCD14546657
  • Cas Number : 774239-27-9
  • Molecular Weight : 227.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0581

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0581
Availability details for
Quantity
  • Availability
  • Add to Cart