COMH4211A9DE Combi-Blocks, Inc.

(R)-[1-(2-Fluoro-benzyl)-piperidin-3-yl]-urea hydrochloride

  • MDL number MFCD31613752
  • Cas Number : 2270918-27-7
  • Molecular Weight : 287.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0582

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0582
Availability details for
Quantity
  • Availability
  • Add to Cart