COMH4211A9DF Combi-Blocks, Inc.

tert-Butyl 4-(6-aminopyridazin-3-yl)piperazine-1-carboxylate

  • MDL number MFCD28127831
  • Cas Number : 1211444-09-5
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0587
Availability details for
Quantity
  • Availability
  • Add to Cart