COMH4211A9E0 Combi-Blocks, Inc.

4-(Cyclopropylmethoxy)-2-fluorobenzonitrile

  • MDL number MFCD23426444
  • Cas Number : 1369882-54-1
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0589
Availability details for
Quantity
  • Availability
  • Add to Cart