COMH4211A9E1 Combi-Blocks, Inc.

Methyl 2-amino-3-(tetrahydro-2h-pyran-4-yl)propanoate

  • MDL number MFCD13193139
  • Cas Number : 1192057-13-8
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0590

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0590
Availability details for
Quantity
  • Availability
  • Add to Cart