COMH4211A9E2 Combi-Blocks, Inc.

4-Bromo-2-isobutoxy-1-nitro-benzene

  • MDL number MFCD30188010
  • Cas Number : 2150940-22-8
  • Molecular Weight : 274.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2-isobutoxy-1-nitrobenzene
Inchi Code 1S/C10H12BrNO3/c1-7(2)6-15-10-5-8(11)3-4-9(10)12(13)14/h3-5,7H,6H2,1-2H3
Inchi Key DYOOAPIKOIGIES-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0591

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0591
Availability details for
Quantity
  • Availability
  • Add to Cart