COMH4211A9E3 Combi-Blocks, Inc.

1-[3-Bromo-4-(propan-2-yloxy)phenyl]ethan-1-amine

  • MDL number MFCD17247745
  • Cas Number : 1344362-49-7
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0593
Availability details for
Quantity
  • Availability
  • Add to Cart